Assay Details
| Gene Symbol: Teddm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178244.3 | NP_839975.2 | 1 | 124 | 89 | |
| GenBank mRNA | CJ048231.1 | - | 1 | 530 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_178244.3 transcripts.
Set Membership: |
|

Teddm1a
Genomic Map