Assay Details
| Gene Symbol: Lrrc4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198250.1 | NP_937893.1 | 1 - 2 | 44 | 68 | |
| GenBank mRNA | BC060263.1 | - | 1 - 2 | 44 | 5686281 | 68 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC060263.1, NM_198250.1 transcripts.
Set Membership: |
|

Lrrc4b
Genomic Map