Assay Details
| Gene Symbol: Lrrc4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198250.1 | NP_937893.1 | 2 - 3 | 381 | 65 | |
| GenBank mRNA | AK140418.1 | - | 1 - 2 | 328 | 65 | |
| BC053486.1 | - | 1 - 2 | 245 | 65 | ||
| BC060263.1 | - | 2 - 3 | 381 | 5686281 | 65 | |
More Information
Set Membership: |
|

Lrrc4b
Genomic Map