Assay Details
| Gene Symbol: Zbtb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198886.3 | NP_942589.3 | 1 - 2 | 205 | 118 | |
| GenBank mRNA | BC020447.1 | - | 1 - 2 | 144 | 118 | |
| BC062159.1 | - | 1 - 2 | 205 | 30135322 | 118 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC062159.1, NM_198886.3 transcripts.
Set Membership: |
|

Zbtb12
Genomic Map