Assay Details
| Gene Symbol: Nr1h5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198658.2 | NP_941060.2 | 2 - 3 | 167 | 96 | |
| NM_201619.2 | NP_963913.2 | 2 - 3 | 167 | 96 | ||
| NM_201622.2 | NP_963916.2 | 2 - 3 | 167 | 96 | ||
| GenBank mRNA | AY094586.1 | - | 2 - 3 | 167 | 96 | |
| AY094587.1 | - | 1 - 2 | 62 | 96 | ||
| AY094588.1 | - | 1 - 2 | 62 | 96 | ||
| AY094589.1 | - | 1 - 2 | 62 | 96 | ||
| AY094590.1 | - | 1 - 2 | 62 | 96 | ||
| AY094591.1 | - | 1 - 2 | 62 | 96 | ||
| BC119524.1 | - | 1 - 2 | 62 | 40056073 | 96 | |
| BC119525.1 | - | 1 - 2 | 62 | 40056074 | 96 | |
More Information
Set Membership: |
|

Nr1h5
Genomic Map