Assay Details
| Gene Symbol: Nr1h5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198658.2 | NP_941060.2 | 8 - 9 | 1130 | 75 | |
| NM_201622.2 | NP_963916.2 | 8 - 9 | 956 | 75 | ||
| GenBank mRNA | AY094586.1 | - | 8 - 9 | 1130 | 75 | |
| AY094587.1 | - | 7 - 8 | 1025 | 75 | ||
| AY094589.1 | - | 7 - 8 | 1025 | 75 | ||
| AY094591.1 | - | 7 - 8 | 851 | 75 | ||
| BC119524.1 | - | 7 - 8 | 1025 | 40056073 | 75 | |
More Information
Set Membership: |
|

Nr1h5
Genomic Map