Assay Details
| Gene Symbol: Osm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013365.2 | NP_001013383.1 | 1 - 2 | 130 | 71 | |
| GenBank mRNA | AK155637.1 | - | 1 - 2 | 81 | 71 | |
| BC099866.1 | - | 1 - 2 | 82 | 40040107 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK155637.1, BC099866.1, NM_001013365.2 transcripts.
Set Membership: |
|

Osm
Genomic Map