Assay Details
| Gene Symbol: Osm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013365.2 | NP_001013383.1 | 2 - 3 | 266 | 90 | |
| GenBank mRNA | AK155637.1 | - | 2 - 3 | 217 | 90 | |
| BC099866.1 | - | 2 - 3 | 218 | 40040107 | 90 | |
More Information
Set Membership: |
|

Osm
Genomic Map