Assay Details
| Gene Symbol: E130308A19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015681.2 | NP_001015681.1 | 2 - 3 | 1472 | 112 | |
| GenBank mRNA | AK047478.1 | - | 2 - 3 | 1473 | 112 | |
| AK081604.1 | - | 2 - 3 | 1356 | 112 | ||
| AK087509.1 | - | 2 - 3 | 1469 | 112 | ||
| BC017649.1 | - | 2 - 3 | 1239 | 4924105 | 112 | |
More Information
Set Membership: |
|

E130308A19Rik
Genomic Map