Assay Details
| Gene Symbol: Zdhhc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168516.1 | NP_001161988.1 | 2 - 3 | 346 | 75 | |
| NM_001168517.1 | NP_001161989.1 | 2 - 3 | 393 | 75 | ||
| NM_027476.3 | NP_081752.2 | 2 - 3 | 639 | 75 | ||
| GenBank mRNA | AK017737.1 | - | 2 - 3 | 604 | 75 | |
| AK028164.1 | - | 2 - 3 | 602 | 75 | ||
| AK043923.1 | - | 2 - 3 | 596 | 75 | ||
| AK169337.1 | - | 2 - 3 | 598 | 75 | ||
| AK170233.1 | - | 2 - 3 | 623 | 75 | ||
| AK170706.1 | - | 2 - 3 | 623 | 75 | ||
| AK171568.1 | - | 2 - 3 | 623 | 75 | ||
| AK172151.1 | - | 2 - 3 | 393 | 75 | ||
| BC056621.1 | - | 2 - 3 | 361 | 75 | ||
| BC071194.1 | - | 2 - 3 | 580 | 75 | ||
| CA559443.1 | - | 2 - 3 | 346 | 75 | ||
| CD354313.1 | - | 2 - 3 | 302 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK172151.1, NM_001168516.1, NM_001168517.1 transcripts.
Set Membership: |
|

Zdhhc24
Genomic Map