Assay Details
| Gene Symbol: Acaa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146230.3 | NP_666342.1 | 2 - 3 | 363 | 95 | |
| GenBank mRNA | AK145953.1 | - | 2 - 3 | 292 | 95 | |
| AK160568.1 | - | 2 - 3 | 210 | 95 | ||
| AY273812.1 | - | 2 - 3 | 361 | 95 | ||
| BC019882.1 | - | 2 - 3 | 276 | 5123887 | 95 | |
More Information
Set Membership: |
|

Acaa1b
Genomic Map