Assay Details
| Gene Symbol: Klrb1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159904.1 | NP_001153376.1 | 6 | 1123 | 92 | |
| NM_008527.2 | NP_032553.2 | 6 | 1114 | 92 | ||
| GenBank mRNA | AK079743.1 | - | 6 | 1125 | 92 | |
| AK136745.1 | - | 1 | 56 | 92 | ||
| DQ237929.1 | - | 6 | 960 | 92 | ||
| M77678.1 | - | 6 | 93 | |||
More Information
Set Membership: |
|

Klrb1c
Genomic Map