Assay Details
| Gene Symbol: Tmem174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026685.2 | NP_080961.1 | 2 | 1421 | 107 | |
| GenBank mRNA | AK057630.1 | - | 2 | 1355 | 107 | |
| BC030878.1 | - | 2 | 1376 | 4241631 | 107 | |
More Information
Set Membership: |
|

Tmem174
Genomic Map