Assay Details
| Gene Symbol: Ccl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011332.3 | NP_035462.2 | 3 | 273 | 59 | |
| GenBank mRNA | AF125570.1 | - | 3 | 362 | 59 | |
| AF125571.1 | - | 3 | 304 | 59 | ||
| AF125572.1 | - | 3 | 277 | 59 | ||
| AJ242587.1 | - | 3 | 252 | 59 | ||
| AK007663.1 | - | 3 | 274 | 59 | ||
| BC028505.1 | - | 3 | 257 | 3328278 | 59 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P114462 | AJ242587.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P114462 | AJ242587.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286031 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78528 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 71222 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157627 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P114462 | AJ242587.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P114462 | AJ242587.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P114462 | AJ242587.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P114462 | AJ242587.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26079565 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157627 | - | - |
| Whole Mouse Genome G4122A | A_51_P114462 | AJ242587.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P114462 | AJ242587.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133633 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157627 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06896982 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06896985 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06896987 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06896998 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06896979 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06896989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26079578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07178717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06811484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06896987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06860791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07078328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07091947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06896979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06811487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07444471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06805673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07018278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06805672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06805674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06896998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07349432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07319242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06897149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06805676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07018277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07319252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26079521 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10574227 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574229 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574228 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419413_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10574226 | - | - |
| Mouse Genome 430 2.0 Array | 1419413_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419413_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20295_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20295_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800008741.mm.1 | NM_011332.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008741.mm.1 | NM_011332.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008741.mm.1 | TC0800001121.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008739.mm.1 | NM_011332.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008739.mm.1 | NM_011332.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008739.mm.1 | TC0800001121.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008740.mm.1 | NM_011332.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008740.mm.1 | NM_011332.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008740.mm.1 | TC0800001121.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419413_PM_at | - | - |
| Mouse Expression Set 430 | 1419413_at | - | - |
| Murine Genome U74v2 Set | 97783_PM_at | - | - |
| Murine Genome U74v2 Set | 97783_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419413_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419413_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419413_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419413_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 913884 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 630121 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 630121 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000630121 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2717015 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2717015 | - | - |
| mouse-6 v1.1 (Illumina) | scl33474.3.1_12-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 630121 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33474.3.1_12-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 630121 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2717015 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2717015 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl33474.3.1_12-S | - | - |

Ccl17
Genomic Map