Assay Details
| Gene Symbol: TMUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015095447.2 | XP_014950933.1 | 1 - 2 | 1217 | 67 | |
| XM_027969020.1 | XP_027824821.1 | 2 - 3 | 516 | 67 | ||
More Information
Set Membership: |
|

TMUB1
Genomic Map