Assay Details
| Gene Symbol: HSD17B10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022064.3 | XP_004022113.1 | 2 - 3 | 254 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSD17B10| Gene Symbol: HSD17B10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022064.3 | XP_004022113.1 | 2 - 3 | 254 | 57 | |
Set Membership: |
|