Assay Details
Gene Symbol: SAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021430.4 | XP_004021479.2 | 5 - 6 | 479 | 76 | |
XM_004021431.4 | XP_004021480.2 | 6 - 7 | 676 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
