Assay Details
| Gene Symbol: TMEM143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100630.2 | XP_014956116.2 | 6 - 7 | 1090 | 61 | |
| XM_015100631.2 | XP_014956117.2 | 5 - 6 | 894 | 61 | ||
| XM_027978557.1 | XP_027834358.1 | 6 - 7 | 1099 | 61 | ||
More Information
Set Membership: |
|

TMEM143
Genomic Map