Assay Details
| Gene Symbol: RAB3IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094745.2 | XP_014950231.1 | 8 - 9 | 1368 | 70 | |
| XM_015094746.2 | XP_014950232.1 | 8 - 9 | 1618 | 70 | ||
| XM_027967461.1 | XP_027823262.1 | 8 - 9 | 1271 | 70 | ||
| XM_027967462.1 | XP_027823263.1 | 7 - 8 | 1522 | 70 | ||
More Information
Set Membership: |
|

RAB3IP
Genomic Map