Assay Details
Gene Symbol: FAM107B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012188404.3 | XP_012043794.1 | 2 - 3 | 389 | 85 | |
XM_027976651.1 | XP_027832452.1 | 2 - 3 | 254 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
