Assay Details
| Gene Symbol: LIN37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015175.4 | XP_004015224.1 | 1 - 2 | 210 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LIN37| Gene Symbol: LIN37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015175.4 | XP_004015224.1 | 1 - 2 | 210 | 54 | |
Set Membership: |
|