Assay Details
| Gene Symbol: RNF126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970131.1 | XP_027825932.1 | 5 - 6 | 587 | 65 | |
| XM_027970132.1 | XP_027825933.1 | 5 - 6 | 545 | 65 | ||
| XM_027970133.1 | XP_027825934.1 | 5 - 6 | 496 | 65 | ||
More Information
Set Membership: |
|

RNF126
Genomic Map