Assay Details
Gene Symbol: GSKIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957375.1 | XP_027813176.1 | 2 - 3 | 321 | 59 | |
XM_027957376.1 | XP_027813177.1 | 2 - 3 | 927 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
