Assay Details
| Gene Symbol: BTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971079.1 | XP_027826880.1 | 3 - 4 | 591 | 74 | |
| XM_027971080.1 | XP_027826881.1 | 3 - 4 | 591 | 74 | ||
| XM_027971081.1 | XP_027826882.1 | 2 - 3 | 245 | 74 | ||
More Information
Set Membership: |
|

BTC
Genomic Map