Assay Details
Gene Symbol: DDX24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017966.3 | XP_004018015.2 | 4 - 5 | 1480 | 60 | |
XM_012098978.2 | XP_011954368.2 | 4 - 5 | 1471 | 60 | ||
XM_027957350.1 | XP_027813151.1 | 4 - 5 | 1442 | 60 | ||
XM_027957351.1 | XP_027813152.1 | 4 - 5 | 1434 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
