Assay Details
| Gene Symbol: ATXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130936.1 | NP_001124408.1 | 8 - 9 | 771 | 72 | |
| XM_012098272.3 | XP_011953662.2 | 8 - 9 | 828 | 72 | ||
| GenBank mRNA | EU797603.1 | - | 8 - 9 | 771 | 72 | |
| FJ943989.1 | - | 8 - 9 | 802 | 72 | ||
More Information
Set Membership: |
|

ATXN3
Genomic Map