Assay Details
| Gene Symbol: TUBB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014431.4 | XP_004014480.2 | 2 - 3 | 263 | 79 | |
| XM_027976992.1 | XP_027832793.1 | 2 - 3 | 264 | 79 | ||
More Information
Set Membership: |
|

TUBB1
Genomic Map