Assay Details
| Gene Symbol: SPESP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318078.1 | NP_001305007.1 | 1 - 2 | 72 | 66 | |
| GenBank mRNA | KR003078.1 | - | 1 - 2 | 72 | 66 | |
More Information
Set Membership: |
|

SPESP1
Genomic Map