Assay Details
| Gene Symbol: RIMS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106870.3 | XP_011962260.1 | 8 - 9 | 1175 | 54 | |
| XM_012106921.3 | XP_011962311.1 | 8 - 9 | 1178 | 54 | ||
More Information
Set Membership: |
|

RIMS3
Genomic Map