Assay Details
| Gene Symbol: UBE2R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004140.4 | XP_004004189.1 | 3 - 4 | 831 | 83 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBE2R2| Gene Symbol: UBE2R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004140.4 | XP_004004189.1 | 3 - 4 | 831 | 83 | |