Assay Details
| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977469.1 | XP_027833270.1 | 3 - 4 | 310 | 59 | |
| XM_027977478.1 | XP_027833279.1 | 3 - 4 | 310 | 59 | ||
| XM_027977487.1 | XP_027833288.1 | 2 - 3 | 289 | 59 | ||
More Information
Set Membership: |
|

FCRLB
Genomic Map