Assay Details
| Gene Symbol: ST8SIA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960733.1 | XP_027816534.1 | 7 - 8 | 777 | 56 | |
| XM_027960734.1 | XP_027816535.1 | 6 - 7 | 669 | 56 | ||
More Information
Set Membership: |
|

ST8SIA5
Genomic Map