Assay Details
| Gene Symbol: PATZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017496.4 | XP_004017545.2 | 2 - 3 | 1351 | 69 | |
| XM_027956813.1 | XP_027812614.1 | 2 - 3 | 1351 | 69 | ||
| XM_027956814.1 | XP_027812615.1 | 2 - 3 | 1351 | 69 | ||
More Information
Set Membership: |
|

PATZ1
Genomic Map