Assay Details
| Gene Symbol: TMEM150A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005874.3 | XP_004005923.3 | 5 - 6 | 450 | 61 | |
| XM_027966734.1 | XP_027822535.1 | 5 - 6 | 594 | 61 | ||
| XM_027966735.1 | XP_027822536.1 | 5 - 6 | 619 | 61 | ||
| XM_027966736.1 | XP_027822537.1 | 5 - 6 | 604 | 61 | ||
More Information
Set Membership: |
|

TMEM150A
Genomic Map