Assay Details
| Gene Symbol: ABHD17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964503.1 | XP_027820304.1 | 2 - 3 | 669 | 101 | |
| XM_027964504.1 | XP_027820305.1 | 2 - 3 | 1074 | 101 | ||
| XM_027964505.1 | XP_027820306.1 | 2 - 3 | 1728 | 101 | ||
| XM_027964506.1 | XP_027820307.1 | 3 - 4 | 673 | 101 | ||
More Information
Set Membership: |
|

ABHD17B
Genomic Map