Assay Details
| Gene Symbol: ETV5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957523.1 | XP_027813324.1 | 11 - 12 | 1414 | 69 | |
| XM_027957527.1 | XP_027813328.1 | 11 - 12 | 1411 | 69 | ||
More Information
Set Membership: |
|

ETV5
Genomic Map