Assay Details
| Gene Symbol: TLR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048231.1 | NP_001041696.1 | 1 | 2320 | 64 | |
| GenBank mRNA | AM981300.1 | - | 1 | 2321 | 64 | |
| DQ890157.1 | - | 1 | 2320 | 64 | ||
More Information
Set Membership: |
|

TLR2
Genomic Map