Assay Details
| Gene Symbol: RILPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161873.1 | NP_001155345.1 | 2 - 3 | 513 | 62 | |
| XM_027980227.1 | XP_027836028.1 | 2 - 3 | 778 | 62 | ||
| GenBank mRNA | FJ943981.1 | - | 2 - 3 | 513 | 62 | |
More Information
Set Membership: |
|

RILPL2
Genomic Map