Assay Details
| Gene Symbol: METTL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267877.1 | NP_001254806.1 | 6 - 7 | 1170 | 66 | |
| GenBank mRNA | JQ348899.1 | - | 6 - 7 | 1170 | 66 | |
More Information
Set Membership: |
|

METTL4
Genomic Map