Assay Details
| Gene Symbol: HSD17B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267882.1 | NP_001254811.1 | 3 - 4 | 721 | 59 | |
| GenBank mRNA | JQ769145.1 | - | 3 - 4 | 721 | 59 | |
More Information
Set Membership: |
|

HSD17B2
Genomic Map