Assay Details
| Gene Symbol: PITX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178053.1 | NP_001171524.1 | 3 - 4 | 410 | 59 | |
| XM_027960148.1 | XP_027815949.1 | 1 - 2 | 4472 | 59 | ||
| XM_027960149.1 | XP_027815950.1 | 2 - 3 | 338 | 59 | ||
| GenBank mRNA | FN432137.1 | - | 3 - 4 | 410 | 59 | |
| FN432138.1 | - | 3 - 4 | 301 | 59 | ||
| FN432139.1 | - | 2 - 3 | 290 | 59 | ||
More Information
Set Membership: |
|

PITX3
Genomic Map