Assay Details
| Gene Symbol: MB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078658.1 | NP_001072126.1 | 2 - 3 | 312 | 97 | |
| XM_012159955.3 | XP_012015345.1 | 4 - 5 | 473 | 97 | ||
| XM_027965775.1 | XP_027821576.1 | 3 - 4 | 442 | 97 | ||
| GenBank mRNA | DQ995220.1 | - | 2 - 3 | 312 | 97 | |

MB
Genomic Map