Assay Details
| Gene Symbol: CYP1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129905.1 | NP_001123377.1 | 6 - 7 | 1376 | 67 | |
| XM_027956968.1 | XP_027812769.1 | 6 - 7 | 1373 | 67 | ||
| XM_027956969.1 | XP_027812770.1 | 6 - 7 | 1370 | 67 | ||
| GenBank mRNA | S79795.1 | - | 6 - 7 | 1376 | 67 | |
More Information
Set Membership: |
|

CYP1A1
Genomic Map