Assay Details
| Gene Symbol: DCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009218.1 | NP_001009218.1 | 2 - 3 | 329 | 64 | |
| XM_012159321.3 | XP_012014711.1 | 2 - 3 | 408 | 64 | ||
| GenBank mRNA | AF125041.1 | - | 2 - 3 | 329 | 64 | |
More Information
Set Membership: |
|

DCN
Genomic Map