Assay Details
| Gene Symbol: LPAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009332.1 | NP_001009332.1 | 2 - 3 | 258 | 59 | |
| XM_012111055.3 | XP_011966445.1 | 2 - 3 | 356 | 59 | ||
| XM_012111061.3 | XP_011966451.1 | 2 - 3 | 335 | 59 | ||
| XM_027963887.1 | XP_027819688.1 | 2 - 3 | 242 | 59 | ||
| GenBank mRNA | U18405.1 | - | 2 - 3 | 258 | 59 | |
More Information
Set Membership: |
|

LPAR1
Genomic Map