Assay Details
| Gene Symbol: SERPINA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100920.2 | NP_001094390.1 | 2 - 3 | 678 | 74 | |
| XM_027962771.1 | XP_027818572.1 | 2 - 3 | 672 | 74 | ||
| GenBank mRNA | X69795.1 | - | 681 | 74 | ||
More Information
Set Membership: |
|

SERPINA7
Genomic Map