Assay Details
| Gene Symbol: FCGR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139452.1 | NP_001132924.1 | 5 - 6 | 860 | 82 | |
| GenBank mRNA | EU589388.1 | - | 5 - 6 | 860 | 82 | |
More Information
Set Membership: |
|

FCGR1A
Genomic Map