Assay Details
Gene Symbol: EPCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162564.1 | NP_001156036.1 | 2 - 3 | 392 | 68 | |
GenBank mRNA | FJ958360.1 | - | 2 - 3 | 392 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
