Assay Details
| Gene Symbol: FSHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009289.1 | NP_001009289.1 | 4 - 5 | 495 | 64 | |
| XM_027965759.1 | XP_027821560.1 | 4 - 5 | 633 | 64 | ||
| GenBank mRNA | L07302.1 | - | 4 - 5 | 495 | 64 | |
| L12767.1 | - | 4 - 5 | 414 | 64 | ||
| L36115.1 | - | 4 - 5 | 495 | 64 | ||
More Information
Set Membership: |
|

FSHR
Genomic Map