Assay Details
| Gene Symbol: NPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009452.1 | NP_001009452.1 | 3 - 4 | 345 | 89 | |
| GenBank mRNA | AJ417904.1 | - | 3 - 4 | 345 | 89 | |
More Information
Set Membership: |
|

NPY
Genomic Map